PRIH11AAF1E5 Princeton BioMolecular Research, Inc.

2-(5-Amino-[1,3,4]thiadiazol-2-ylsulfanylmethyl)-5-hydroxy-pyran-4-one

  • MDL number MFCD11618414
  • Cas Number : 1094533-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart