PRIH11AAF1F4 Princeton BioMolecular Research, Inc.

5-Bromo-1,4-dimethyl-1H-pyridin-2-one

  • MDL number MFCD22688548
  • Cas Number : 889865-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart