PRIH11AAF202 Princeton BioMolecular Research, Inc.

4-(2,5-Dimethyl-phenyl)-benzo[d][1,2]oxazin-1-one

  • MDL number MFCD26097375
  • Cas Number : 2224-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart