PRIH11AAF20B Princeton BioMolecular Research, Inc.

3-Hydroxyimino-cyclopentanecarboxylic acid

  • MDL number MFCD19228333
  • Cas Number : 6300-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart