PRIH11AAF21E Princeton BioMolecular Research, Inc.

2-Mercapto-5-(4-methoxy-phenyl)-3-phenyl-3H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD01990252
  • Cas Number : 296899-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart