PRIH11AAF225 Princeton BioMolecular Research, Inc.

2-Carboxymethoxy-5-chloro-benzoic acid methyl ester

  • MDL number MFCD12033963
  • Cas Number : 21456-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart