PRIH11AAF229 Princeton BioMolecular Research, Inc.

2-(4-Bromo-butoxy)-phenol

  • MDL number MFCD08447796
  • Cas Number : 16837-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart