PRIH11AAF22B Princeton BioMolecular Research, Inc.

Tetrahydro-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD19668134
  • Cas Number : 113990-87-7
  • Molecular Weight : 146.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart