PRIH11AAF233 Princeton BioMolecular Research, Inc.

2-Amino-6-hydroxymethyl-8-oxo-4-phenyl-4,8-dihydro-pyrano[3,2-b]pyran-3-carbonitrile

  • MDL number MFCD07187842
  • Cas Number : 194282-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart