PRIH11AAF234 Princeton BioMolecular Research, Inc.

7-Hydroxy-4,5-dimethyl-chromen-2-one

  • MDL number MFCD01935666
  • Cas Number : 51786-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart