PRIH11AAF238 Princeton BioMolecular Research, Inc.

4-(Pyrrolidine-1-sulfonyl)-benzoic acid methyl ester

  • MDL number MFCD02573093
  • Cas Number : 333787-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart