PRIH11AAF23C Princeton BioMolecular Research, Inc.

11-Amino-undecanoic acid methyl ester

  • MDL number MFCD09923522
  • Cas Number : 28691-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart