PRIH11AAF240 Princeton BioMolecular Research, Inc.

2,2,4,7-Tetramethyl-1,2-dihydro-quinoline

  • MDL number MFCD00623490
  • Cas Number : 1810-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart