PRIH11AAF245 Princeton BioMolecular Research, Inc.

N-[2-(5-Amino-4-ethyl-2-methoxy-phenoxy)-ethyl]-acetamide

  • MDL number MFCD28155446
  • Cas Number : 1797979-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart