PRIH11AAF259 Princeton BioMolecular Research, Inc.

4,7-Dichloro-5-hydroxy-benzo[1,3]oxathiol-2-one

  • MDL number MFCD00744253
  • Cas Number : 7735-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart