PRIH11AAF26A Princeton BioMolecular Research, Inc.

1-tert-butyl 6-methyl 3,4-dihydroquinoline-1,6(2H)-dicarboxylate

  • MDL number MFCD29910134
  • Cas Number : 1398413-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart