PRIH11AAF26B Princeton BioMolecular Research, Inc.

4-(5-methyl-2-pyridin-2-yl-1H-indol-3-yl)butanoic acid

  • MDL number MFCD06254713
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart