PRIH11AAF26F Princeton BioMolecular Research, Inc.

3,4,5-trimethyl-2-furoic acid

  • MDL number MFCD17069367
  • Cas Number : 34289-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart