PRIH11AAF272 Princeton BioMolecular Research, Inc.

(6E)-6-(hydroxyimino)bicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD30378841
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart