PRIH11AAF276 Princeton BioMolecular Research, Inc.

3-(4-bromo-3-methylisoxazol-5-yl)propanoic acid

  • MDL number MFCD30830566
  • Molecular Weight : 234.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart