PRIH11AAF27C Princeton BioMolecular Research, Inc.

5-ethyl-4-methyl-4H-1,2,4-triazol-3-amine

  • MDL number MFCD03731311
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart