PRIH11AAF28D Princeton BioMolecular Research, Inc.

4-(2,6,6-trimethyl-4-oxo-4,5,6,7-tetrahydro-1H-indol-1-yl)benzenesulfonamide

  • MDL number MFCD09250782
  • Molecular Weight : 332.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart