PRIH11AAF28F Princeton BioMolecular Research, Inc.

2-(1-methyl-2-oxopropyl)cyclohexane-1,3-dione

  • MDL number MFCD09837232
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart