PRIH11AAF293 Princeton BioMolecular Research, Inc.

6-fluoro-2-(morpholin-4-ylcarbonyl)-2,3-dihydro-4H-chromen-4-one

  • MDL number MFCD09064466
  • Molecular Weight : 279.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart