PRIH11AAF2A3 Princeton BioMolecular Research, Inc.

3,5-dimethylphenyl 4,7,7-trimethyl-2,3-dioxobicyclo[2.2.1]heptane-1-carboxylate

  • MDL number MFCD04066552
  • Molecular Weight : 314.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart