PRIH11AAF2A7 Princeton BioMolecular Research, Inc.

6-bromo-2-methoxy-1-(methoxymethyl)naphthalene

  • MDL number MFCD03468991
  • Molecular Weight : 281.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart