PRIH11AAF2AB Princeton BioMolecular Research, Inc.

N-(4-iodophenyl)-4,7,7-trimethyl-3-oxobicyclo[2.2.1]heptane-1-carboxamide

  • MDL number MFCD03659918
  • Molecular Weight : 397.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart