PRIH11AAF2B7 Princeton BioMolecular Research, Inc.

4-[3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl]benzoic acid

  • MDL number MFCD03731314
  • Cas Number : 884239-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart