PRIH11AAF2BA Princeton BioMolecular Research, Inc.

(4Z)-6,6-dimethyl-6,7-dihydro-1-benzofuran-4(5H)-one thiosemicarbazone

  • MDL number MFCD03011703
  • Molecular Weight : 237.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart