PRIH11AAF2BD Princeton BioMolecular Research, Inc.

2-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD02918344
  • Cas Number : 17969-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart