PRIH11AAF2BE Princeton BioMolecular Research, Inc.

(4Z)-3,6,6-trimethyl-1-phenyl-1,5,6,7-tetrahydro-4H-indazol-4-one oxime

  • MDL number MFCD06496301
  • Molecular Weight : 269.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart