PRIH11AAF2C3 Princeton BioMolecular Research, Inc.

4-ethoxyquinazoline

  • MDL number MFCD01646083
  • Cas Number : 16347-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart