PRIH11AAF2CB Princeton BioMolecular Research, Inc.

(2E)-bicyclo[2.2.1]heptan-2-one oxime

  • MDL number MFCD09836262
  • Cas Number : 4576-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart