PRIH11AAF2CC Princeton BioMolecular Research, Inc.

1-(2-chloro-1,1-dimethylethyl)-4-ethoxybenzene

  • MDL number MFCD03731270
  • Cas Number : 83493-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart