PRIH11AAF311 Princeton BioMolecular Research, Inc.

5-chloro-2-(methylthio)-N-pyridin-3-ylpyrimidine-4-carboxamide

  • MDL number MFCD06630135
  • Molecular Weight : 280.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart