PRIH11AAF326 Princeton BioMolecular Research, Inc.

5-chloro-2-(methylthio)pyrimidine-4-carboxamide

  • MDL number MFCD08744823
  • Cas Number : 902243-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart