PRIH11AAF32A Princeton BioMolecular Research, Inc.

[(2-bromopyridin-3-yl)oxy]acetic acid

  • MDL number MFCD09837166
  • Cas Number : 118650-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart