PRIH11AAF32F Princeton BioMolecular Research, Inc.

3-methyl-5-[(E)-2-phenylvinyl]isoxazol-4-amine

  • MDL number MFCD22201511
  • Cas Number : 51978-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart