PRIH11AAF336 Princeton BioMolecular Research, Inc.

[1-(3-methylquinoxalin-2-yl)pyrrolidin-3-yl]methanol

  • MDL number MFCD17966973
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart