PRIH28C053FA Princeton BioMolecular Research, Inc.

4,5-Dibromo-3-(2-methoxy-ethoxy)-thiophene-2-carboxylic acid

  • MDL number MFCD26129626
  • Molecular Weight : 360.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart