PRIH3031267B Princeton BioMolecular Research, Inc.

(2-Cyclopropyl-ethyl)-methyl-amine

  • MDL number MFCD13249717
  • Cas Number : 161353-92-0
  • Molecular Weight : 99.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart