PRIH303126CB Princeton BioMolecular Research, Inc.

cyclopentyl(pyridin-3-yl)acetic acid

  • MDL number MFCD24334874
  • Cas Number : 1515051-83-8
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart