PRIH303127C2 Princeton BioMolecular Research, Inc.

3-Hydroxy-1-methyl-4-pyridin-3-yl-1H-quinolin-2-one

  • MDL number MFCD30836088
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart