PRIH30329BD9 Princeton BioMolecular Research, Inc.

7-Hydroxy-5-pyridin-4-yl-pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD26382468
  • Cas Number : 1707372-70-0
  • Molecular Weight : 256.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart