PRIH30329BDA Princeton BioMolecular Research, Inc.

7-Hydroxy-5-trifluoromethyl-pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD19312913
  • Cas Number : 1310308-29-2
  • Molecular Weight : 247.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart