PRIH303770CA Princeton BioMolecular Research, Inc.

4-Amino-2-morpholin-4-yl-thiazole-5-carboxylic acid amide

  • MDL number MFCD17780296
  • Cas Number : 99967-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart