PRIH303770CB Princeton BioMolecular Research, Inc.

1-(4-Bromo-phenyl)-ethane-1,2-diamine

  • MDL number MFCD12190313
  • Cas Number : 735208-91-0
  • Molecular Weight : 215.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart