PRIH303770CD Princeton BioMolecular Research, Inc.

6-(2,5-Dimethoxy-phenyl)-3-methyl-imidazo[2,1-b]thiazole-2-carboxylic acid

  • MDL number MFCD11221834
  • Molecular Weight : 318.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart