PRIH303770D0 Princeton BioMolecular Research, Inc.

1-Isobutyl-2-methyl-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid

  • MDL number MFCD28246872
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart