PRIH303770D1 Princeton BioMolecular Research, Inc.

1-Aminomethyl-cyclopropanecarbonitrile

  • MDL number MFCD19204888
  • Cas Number : 1124213-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart